Learn More
Sigma Aldrich Fine Chemicals Biosciences Cas9 mRNA1EA
Catalog No. 502905750
Encompass
Cas9 mRNA generated through in-vitro transcription which has been capped and polyA-tailed. This mRNA should be used in conjunction with purified guideRNA. The use of RNA vs. plasmid constructs avoids complications with promoter-embryo compatibility as well as the possibility of random integration of nuclease and gRNA-expressing plasmids into the host animal genome.
Catalog No. 50-290-5750 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. CAS9MRNA1EA
Cas9 mRNA generated through in-vitro transcription which has been capped and polyA-tailed. This mRNA should be used in conjunction with purified guideRNA. The use of RNA vs. plasmid constructs avoids complications with promoter-embryo compatibility as well as the possibility of random integration of nuclease and gRNA-expressing plasmids into the host animal genome.