Learn More
Cayman Chemical Cas9 mRNACap-1mo5U 100ug
SDP
Catalog No. 502945620
Encompass_Preferred
An mRNA for the expression of Cas9; encapsulation in LNPs can be used for mRNA delivery and expression of Cas9 in vitro or in vivo
Catalog No. 50-294-5620 Supplier Cayman Chemical Supplier No. 41964100 UG
An mRNA for the expression of Cas9; encapsulation in LNPs can be used for mRNA delivery and expression of Cas9 in vitro or in vivo