Learn More
New England Biolabs, Inc. Cas9 Nuclease, S. pyogenes – 2000 pmol
SDP
Catalog No. NC0894039
Encompass
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.
Catalog No. NC0894039 Supplier New England Biolabs, Inc. Supplier No. M0386M
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.