Learn More
INTACT GENOMICS INC Cas9 Nuclease 40 µg
Catalog No. 5000082992
Encompass
Intact Genomics Cas9 Nuclease is the purified recombinant Streptococcus pyogenes Cas9 enzyme containing a nuclear localization signal (NLS) at the C-terminal for targeting to the nucleus. This enzyme is designed to perform CRISPR/Cas9-mediated genome editing (1, 2). The physical purity of this enzyme is ≥98% as assessed by SDS-PAGE with Coomassie blue staining. Product Source:E. coli BL21 (DE3) strain expressing a Cas9 gene from Streptococcus pyogenes with an N-terminal 6xHis tag and C-terminal SV40 nuclear localization signal (NLS).Product Includes: Cas9 Nuclease 10x Cas9 Nuclease Reaction Buffer
Catalog No. 50-000-82992 Supplier INTACT GENOMICS INC Supplier No. 3270
Intact Genomics Cas9 Nuclease is the purified recombinant Streptococcus pyogenes Cas9 enzyme containing a nuclear localization signal (NLS) at the C-terminal for targeting to the nucleus. This enzyme is designed to perform CRISPR/Cas9-mediated genome editing (1, 2). The physical purity of this enzyme is ≥98% as assessed by SDS-PAGE with Coomassie blue staining. Product Source:E. coli BL21 (DE3) strain expressing a Cas9 gene from Streptococcus pyogenes with an N-terminal 6xHis tag and C-terminal SV40 nuclear localization signal (NLS).Product Includes: Cas9 Nuclease 10x Cas9 Nuclease Reaction Buffer