Learn More
New England Biolabs, Inc. Cas9 Nuclease, S. pyogenes – 70 pmol
SDP
Catalog No. 50607649
Encompass
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.
Catalog No. 50-607-649 Supplier New England Biolabs, Inc. Supplier No. M0386S
 Restricted Item
Add to Cart
Add to Cart
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.