Learn More
ABclonal Technology CAS9 Rabbit pAb
SDP
Supplier:  ABclonal Technology A14997500UL
Encompass_Preferred
Staphylococcus aureus Cas9 (SaCas9) is an RNA-guided endonuclease that targets complementary DNA adjacent to a protospacer adjacent motif (PAM) for cleavage. Its small size facilitates in vivo delivery for genome editing in various organisms.
Catalog No. 50-001-05434