Learn More
Bioworld Casamino Acids, 100 g
Catalog No. 50255675
Encompass_Preferred
Acid hydrolyzed casein with low sodium chloride and iron concentrations. Used in diptheria toxin production. Also known as Casein Acid, Hydrolysate.Appearance: SolidPH Value: 6.6-7.5RTECS Number: FI3519450
Catalog No. 50-255-675 Supplier Bioworld Supplier No. 403000481
Acid hydrolyzed casein with low sodium chloride and iron concentrations. Used in diptheria toxin production. Also known as Casein Acid, Hydrolysate.Appearance: SolidPH Value: 6.6-7.5RTECS Number: FI3519450