Learn More
Research Products International Corp RPI Casamino Acids [Casein acid hydrolysate], 10 Kilograms
SDP
Catalog No. 501976874
Encompass_Preferred
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.
Catalog No. 50-197-6874 Supplier Research Products International Corp Supplier No. C4500010000.0
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.