Learn More
Research Products International Corp Casamino Acids [Casein acid hydrolysate], 1 Kilogram
SDP
Catalog No. 50995824
Encompass_Preferred
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.
Catalog No. 50-995-824 Supplier Research Products International Corp Supplier No. C450001000.0
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.