Learn More
AG Scientific Inc Casamino Acids, 500 G
Supplier:  AG Scientific Inc C2733500GM
Encompass_Preferred
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.CAS Number: 91079-40-2Solubility: WaterStorage Temperature: Room TemperatureResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-7064