Learn More
Research Products International Corp Casamino Acids [Casein acid hydrolysate], 500 Grams
SDP
Catalog No. 50488602
Encompass_Preferred
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.
Catalog No. 50-488-602 Supplier Research Products International Corp Supplier No. C45000500.0
Casein Acid Hydrolysate is a hydrochloric acid digest of casein that contains small amounts of cystine and Tryptophan. All vitamins have been eliminated by the acidification treatment. Provides a source of amino acids as nutrients for growing various organisms tolerant of high salt conditions.