Learn More
Alkali Scientific Casamino Acids [Casein acid hydrolysate], 10 Kg
SDP
Catalog No. 5000661523
Encompass_Preferred
Casamino Acids (Casein Acid Hydrolysate) in this 10 kg bulk format is used as a high-quality nutrient source for microbial growth in a variety of laboratory applications. The hydrolysate provides essential amino acids and peptides that support the growth of a wide range of microorganisms, including bacteria, fungi, and yeasts. The bulk packaging is ideal for high-throughput laboratories, research facilities, and industrial applications that require large quantities of growth media components. Casamino acids are frequently used in microbiological culture media for the cultivation of fastidious organisms that need additional nutrients to grow effectively.
Catalog No. 50-006-61523 Supplier Alkali Scientific Supplier No. ASA002CP
Casamino Acids (Casein Acid Hydrolysate) in this 10 kg bulk format is used as a high-quality nutrient source for microbial growth in a variety of laboratory applications. The hydrolysate provides essential amino acids and peptides that support the growth of a wide range of microorganisms, including bacteria, fungi, and yeasts. The bulk packaging is ideal for high-throughput laboratories, research facilities, and industrial applications that require large quantities of growth media components. Casamino acids are frequently used in microbiological culture media for the cultivation of fastidious organisms that need additional nutrients to grow effectively.