Learn More
Alkali Scientific Casamino Acids [Casein acid hydrolysate], 500 G
SDP
Catalog No. 5000661527
Encompass_Preferred
Casamino Acids (Casein Acid Hydrolysate) is a nutrient-rich powder used in microbiological media to support the growth of bacteria, fungi, and other microorganisms. This 500g pack is ideal for small to medium-scale laboratory experiments and testing. Casamino acids provide essential amino acids and peptides that are required for microbial growth, especially for fastidious organisms that cannot synthesize their own amino acids. This reagent is commonly used in the preparation of growth media for research, quality control, and clinical diagnostics. It is an essential component in culture media formulations for the cultivation of a wide range of microbes.
Catalog No. 50-006-61527 Supplier Alkali Scientific Supplier No. ASA004CP
Casamino Acids (Casein Acid Hydrolysate) is a nutrient-rich powder used in microbiological media to support the growth of bacteria, fungi, and other microorganisms. This 500g pack is ideal for small to medium-scale laboratory experiments and testing. Casamino acids provide essential amino acids and peptides that are required for microbial growth, especially for fastidious organisms that cannot synthesize their own amino acids. This reagent is commonly used in the preparation of growth media for research, quality control, and clinical diagnostics. It is an essential component in culture media formulations for the cultivation of a wide range of microbes.