Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Casamino Acids, OmniPur™, Calbiochem™,

Catalog No. 2240500gm Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-405-00GM 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-405-00GM Supplier MilliporeSigma™ Supplier No. 2240500GM
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 65072-00-6
Molecular Formula C21H41N5O11
Molecular Weight (g/mol) 539.58
MDL Number MFCD00130738
InChI Key XZNUGFQTQHRASN-UHFFFAOYNA-N
Synonym Casein
PubChem CID 57397142
IUPAC Name Casein hydrolysate
SMILES *

Specifications

pH 5.5 to 7.0 (at 25°C 2% H2O)
Color White to Beige
Packaging Plastic Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.