Learn More
Apexbio Technology LLC Casanthranol 8024-48-4 1g
SDP
Catalog No. 5000093928
Encompass_Preferred
Casanthranol is a natural mixture of anthranol glycosides derived from the bark of Rhamnus purshiana commonly known as cascara sagrada Casanthranol exerts its biological activity primarily through intestinal bacterial hydrolysis releasing anthraquinone aglycones that interact with colonic mucosa to enhance bowel motility and stimulate fluid accumulation in the intestinal lumen Based on these pharmacological properties casanthranol holds research potential in studies of intestinal motility investigation of laxative mechanisms microbiota-mediated glycoside metabolism and as a chemical probe for elucidating gastrointestinal physiology and anthraquinone glycoside bioactivity
Catalog No. 50-000-93928 Supplier Apexbio Technology LLC Supplier No. A83901000
Casanthranol is a natural mixture of anthranol glycosides derived from the bark of Rhamnus purshiana commonly known as cascara sagrada Casanthranol exerts its biological activity primarily through intestinal bacterial hydrolysis releasing anthraquinone aglycones that interact with colonic mucosa to enhance bowel motility and stimulate fluid accumulation in the intestinal lumen Based on these pharmacological properties casanthranol holds research potential in studies of intestinal motility investigation of laxative mechanisms microbiota-mediated glycoside metabolism and as a chemical probe for elucidating gastrointestinal physiology and anthraquinone glycoside bioactivity