Learn More
Aviva Systems Biology Corp CASC3 Antibody - N-terminal region (ARP54817_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP54817_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CASC3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CASC3. Alternate names: BTZ, MLN51ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61328