Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Cascade Blue™ hydrazide, Trisodium Salt

Catalog No. C687
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C687 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C687 Supplier Invitrogen™ Supplier No. C687
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The cell-impermeant, fixable, polar tracer Cascade Blue™ hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis.

The cell-impermeant, fixable, polar tracer Cascade Blue™ hydrazide can be used to investigate cell-cell and cell-liposome fusion as well as membrane permeability and transport through gap junctions or cell uptake during pinocytosis. This water-soluble tracer can be introduced into cells by whole-cell patch clamping, iontophoresis, osmotic lysis of pinocytic vesicles or comparable methods. Additionally, carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (Cat. No. E2247).

To make a concentrated stock solution, dissolve in H2O up to ∼1% (w/v). Store frozen and protected from light.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 10 mg
Product Type Hydrazide
Molecular Weight (g/mol) 596.44 G/MOL
Content And Storage Store at room temperature and protect from light.
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Shipping Condition Room Temperature
Reactive Moiety Amine, Hydrazide
Label or Dye Cascade Blue™
Label Type Classic Dyes
Product Line CASCADE BLUE
Emission 412 nm
Excitation 399 nm
Show More Show Less
What is the recommended concentration for preparation of Cascade Blue hydrazide, Trisodium Salt (Cat. No. C687) stock solution?

We recommend preparing a 1% w/v stock solution in sterile water. Please note that the dye can aggregate in solution over time.

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.