Learn More
Enterprise Technology Solutions Cascades Wiper Roll, Scrim-Reinforced, 9-3/4"x275', 6RL/CT, WE
SDP
Catalog No. 5000831587
Encompass_Preferred
Made with a high percentage of recycled fiber, Cascades PRO Tuff-Job Scrim Reinforced Wipers offer multi-ply, high wet-strength paper for superior absorbency. They are engineered with a polyester scrim webbing within the plies of the paper to add extra strength and durability for medium-duty cleanups for janitorial, automotive, manufacturing and maintenance needs. These wipes in a nonperforated roll provide a high wipe-dry performance, especially for glass and other smooth surfaces.
Catalog No. 50-008-31587 Supplier Enterprise Technology Solutions Supplier No. CSDW220
Made with a high percentage of recycled fiber, Cascades PRO Tuff-Job Scrim Reinforced Wipers offer multi-ply, high wet-strength paper for superior absorbency. They are engineered with a polyester scrim webbing within the plies of the paper to add extra strength and durability for medium-duty cleanups for janitorial, automotive, manufacturing and maintenance needs. These wipes in a nonperforated roll provide a high wipe-dry performance, especially for glass and other smooth surfaces.