Learn More
Lakeside Manufacturing Inc Closed Case Cart, Two Door, Solid Stainless Steel Shelves, 36-In Shelf Width
Catalog No. NC3136667
Encompass_Preferred
The Lakeside 6937S Closed Case Cart has two doors with solid stainless steel shelving that has a 36" shelf width. This unit is an all-welded stainless steel construction. The hold open latch secures the door open for easy cleaning. Door latch keeps contents safe and sterile (locking model available). The extended push handle offers better control. This model has solid stainless steel pull-out shelves equipped with a locking mechanism. The innovative floor design directs water to the drain area. Made in the USA.
Catalog No. NC3136667 Supplier Lakeside Manufacturing Inc Supplier No. 6937S
Add to Cart
Add to Cart
Request a Quote
The Lakeside 6937S Closed Case Cart has two doors with solid stainless steel shelving that has a 36" shelf width. This unit is an all-welded stainless steel construction. The hold open latch secures the door open for easy cleaning. Door latch keeps contents safe and sterile (locking model available). The extended push handle offers better control. This model has solid stainless steel pull-out shelves equipped with a locking mechanism. The innovative floor design directs water to the drain area. Made in the USA.