Learn More
Research Products International Corp Case Filter Pipet Tips, 200ul, 960/cs, 5/cs
SDP
Supplier:  Research Products International Corp 141062CS
Encompass
Tips are natural, polypropylene, and low binding. Racks are sterile. Pipet tips are Certified RNase/DNase Free and Human DNA Free. Tips are also Certified PCR Inhibitor Free and Pyrogen Free. Holds 1 - 200 ul with graduation lines at 10, 50, and 100 ul.
Catalog No. 50-197-7234