Learn More
Kuhner Shaker Inc CASE OF FOUR EXTRA HIGH SQUARE
Catalog No. 502570147
Encompass
Case of four Extra high square deepwell MTP (polystyrene, 85 mm high 6 wells of 100 ml volume, Pyramid bottom, gamma irradiated) single use (non-autoclavable)
Catalog No. 50-257-0147 Supplier Kuhner Shaker Inc Supplier No. SMCR1406
Case of four Extra high square deepwell MTP (polystyrene, 85 mm high 6 wells of 100 ml volume, Pyramid bottom, gamma irradiated) single use (non-autoclavable)