Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spectrum™ Chemical Casein
SDP

Catalog No. 18621716 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-716 500 g
18-621-717 2.5 kg
18-621-718 12 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-621-716 Supplier Spectrum™ Chemical Supplier No. CA205500GM
Only null left
Add to Cart
Add to Cart

Specifications

Quantity 500 g
Product Type Casein
Form Solid
Color White, Off-White, Yellowish
Melting Point 280°C/536°F
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.