Learn More
Medchemexpress LLC Casein | 9000-71-9 | 97.4% | 2061.96 | C81H125N22O39P | 25G
SDP
Catalog No. 5000235322
Encompass_Preferred
Casein is a bovine phosphoprotein supplied as a powdered research reagent (CAS 9000-71-9). It is used as a substrate for proteases, in kinase and endogenous metabolite studies, and for general laboratory applications requiring casein protein. The material is listed at 97.4% purity and is offered in multiple sizes, including 25 g.
Catalog No. 50-002-35322 Supplier Medchemexpress LLC Supplier No. HY12586525G
Casein is a bovine phosphoprotein supplied as a powdered research reagent (CAS 9000-71-9). It is used as a substrate for proteases, in kinase and endogenous metabolite studies, and for general laboratory applications requiring casein protein. The material is listed at 97.4% purity and is offered in multiple sizes, including 25 g.