Learn More
Medchemexpress LLC Casein | 9000-71-9 | MFCD00081481 | 97.4% | 2061.96 | C81H125N22O39P | 100G
SDP
Catalog No. 5000235723
Encompass_Preferred
Casein is a phosphoprotein powder (CAS 9000-71-9) commonly used as a biochemical substrate, nutritional component, and industrial ingredient. Supplied as a research-grade powder with a reported purity of 97.4%, it is available in multiple pack sizes and accompanied by a certificate of analysis and safety data sheet.
  • Provided as a powder suitable for laboratory and industrial use
  • Reported purity of 97.4%
  • Can be separated into α2-, κ-, β-, and γ-casein components
  • Used as a substrate in enzymatic and kinase assays
  • Enhances calcium absorption and supports dental research applications
  • Applicable for coatings, adhesives, and packaging in paper, leather, and textile industries
  • COA and SDS available for quality and safety verification
Catalog No. 50-002-35723 Supplier Medchemexpress LLC Supplier No. HY125865100G
Casein is a phosphoprotein powder (CAS 9000-71-9) commonly used as a biochemical substrate, nutritional component, and industrial ingredient. Supplied as a research-grade powder with a reported purity of 97.4%, it is available in multiple pack sizes and accompanied by a certificate of analysis and safety data sheet.
  • Provided as a powder suitable for laboratory and industrial use
  • Reported purity of 97.4%
  • Can be separated into α2-, κ-, β-, and γ-casein components
  • Used as a substrate in enzymatic and kinase assays
  • Enhances calcium absorption and supports dental research applications
  • Applicable for coatings, adhesives, and packaging in paper, leather, and textile industries
  • COA and SDS available for quality and safety verification