Learn More
Bioworld Casein Blocking Buffer with Fish Gelatin, 100 mL
Supplier:  Bioworld 104500011
Encompass_Preferred
Casein Fish Gelatin Blocking Buffer is a non-mammalian blocking solution that does not cross-react with mammalian antibodies and is suitable for ELISA assays. Blocking buffer is supplied in a 10X concentration of physiological saline buffer. Technical Information: Casein Blocker with Fish Gelatin is a casein-based blocking agent Includes 3 X 30 mL, 10X concentratePrepares 900ml of working solution1X working solution contains: 1% casein, 0.45% fish gelatin in PBS.
Catalog No. 50-196-446