Learn More
Sigma Aldrich Fine Chemicals Biosciences Casein Blocking Buffer 10x, 500 mL
Catalog No. NC0913654
Encompass
Sigma’s 10x Blocking Buffer is a casein-based blocking agent routinely used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes.
Catalog No. NC0913654 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. B6429500ML
Sigma’s 10x Blocking Buffer is a casein-based blocking agent routinely used to reduce non-specific binding of detection reagents to the Northern and Southern Blot membranes.