Learn More
Bioworld Casein-Blocking Buffer in TBS w/ Azide, 1 L
Catalog No. 50255759
Encompass_Preferred
Product DescriptionCasein-blocking buffer is 3% in TBS with 0.02% sodium azide. It is a liquid used for Western blots and ELISA. Buffer used for blocking excess binding sites on membranes and microplates in antibody-based detection methods.
Catalog No. 50-255-759 Supplier Bioworld Supplier No. 403200322
Product DescriptionCasein-blocking buffer is 3% in TBS with 0.02% sodium azide. It is a liquid used for Western blots and ELISA. Buffer used for blocking excess binding sites on membranes and microplates in antibody-based detection methods.