Learn More
Bioworld Casein-Blocking Buffer in TBS, 500 mL
Supplier:  Bioworld 403200281
Encompass_Preferred
Casein Blocking Buffer in TBS Blot blocking buffer used to block non-specific binding site for antibodies on the membrane. Technical Information: 1% Casein in TBS
Catalog No. 50-255-754