Learn More
Research Products International Corp Casein Blocking Solution in TBS, 1 Liter
SDP
Catalog No. 50488599
Encompass_Preferred
Contains 3% Casein in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.
Catalog No. 50-488-599 Supplier Research Products International Corp Supplier No. C440001000.0
Contains 3% Casein in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.