Learn More
Alkali Scientific Casein Blocking Solution in TBS, 1 L
SDP
Catalog No. 5000661507
Encompass_Preferred
Casein Blocking Solution in TBS (Tris-Buffered Saline) is a ready-to-use reagent designed for protein blocking in laboratory assays. This 1L solution is used to block non-specific binding sites in Western blotting, ELISA, and other immunoassays to ensure accurate detection of target proteins. The casein protein prevents antibodies from binding to surfaces other than the target, improving the specificity and sensitivity of assays. This reagent is particularly useful in protein detection, antibody binding studies, and immunochemistry applications where high precision is required. It is a standard solution in molecular biology and biochemistry laboratories.
Catalog No. 50-006-61507 Supplier Alkali Scientific Supplier No. CAS001CP
Casein Blocking Solution in TBS (Tris-Buffered Saline) is a ready-to-use reagent designed for protein blocking in laboratory assays. This 1L solution is used to block non-specific binding sites in Western blotting, ELISA, and other immunoassays to ensure accurate detection of target proteins. The casein protein prevents antibodies from binding to surfaces other than the target, improving the specificity and sensitivity of assays. This reagent is particularly useful in protein detection, antibody binding studies, and immunochemistry applications where high precision is required. It is a standard solution in molecular biology and biochemistry laboratories.