Learn More
NEOGEN Casein Digest, 500 g
SDP
Catalog No. 502015756
Encompass_Preferred
Casein Digest is an enzymatic digest of casein for use in preparing microbiological culture media in a laboratory setting. Casein Digest is not intended for use in the diagnosis of disease or other conditions in humans.Casein is the main protein of milk and a rich source of amino nitrogen. Casein Digest was developed for use in molecular genetic media formulations, including NZCYM Broth, NZYM Broth, and NZM Broth. The enriched formulations of NZ media are used to cultivate recombinant strains of E. coli and propagating theta bacteriophages.For more information visit https://www.neogen.com/solutions/Microbiology/casein-digest
Catalog No. 50-201-5756 Supplier NEOGEN Supplier No. 700003534
Casein Digest is an enzymatic digest of casein for use in preparing microbiological culture media in a laboratory setting. Casein Digest is not intended for use in the diagnosis of disease or other conditions in humans.Casein is the main protein of milk and a rich source of amino nitrogen. Casein Digest was developed for use in molecular genetic media formulations, including NZCYM Broth, NZYM Broth, and NZM Broth. The enriched formulations of NZ media are used to cultivate recombinant strains of E. coli and propagating theta bacteriophages.For more information visit https://www.neogen.com/solutions/Microbiology/casein-digest