Learn More
Sigma Aldrich Fine Chemicals Biosciences Casein from bovine milk, 9000-71-9, MFCD00081481, 500 G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences C7078500G
Encompass
Technical grade. Casein from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk: α-s1 Casein, α-s2 Casein, β-Casein, and κ-Casein. Casein is proposed to be the main protective constituent in milk.
Catalog No. NC1079502