Learn More
Sigma Aldrich Fine Chemicals Biosciences Casein from bovine milk su5KG
Catalog No. 502903883
Encompass
Casein from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk alpha-s1 Casein alpha-s2 Casein beta-Casein and kappa-Casein. Casein is proposed to be the main protective constituent in milk.
Catalog No. 50-290-3883 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. C58905KG
Casein from bovine milk is a phosphoprotein. There are four main types of Casein which make up approximately 80% of the total protein in bovine milk alpha-s1 Casein alpha-s2 Casein beta-Casein and kappa-Casein. Casein is proposed to be the main protective constituent in milk.