Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Casein Hydrolysate Acid Solution , MP Biomedicals, LLC

Catalog No. MP210476990 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Plastic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP210476990 500 mL Plastic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP210476990 Supplier MP Biomedicals, Inc Supplier No. 0210476990
Add to Cart
Add to Cart

Vitamin free Casein hydrolysate acid solution is a hydrochloric acid digest of casein in which the protein molecule has been substantially reduced to its constituent amino acids

Laboratory media including basal media for vitamin analysis of folic acid, pantothenic acid, riboflavin, thiamin, niacin, and B-12
TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Packaging Plastic
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 0.1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.