Learn More
Eurofins Discoverx Casein Kinase 2α, active, 10 µg. (14-445)
Supplier:  Eurofins Discoverx 14445
Encompass
6xHis-tagged recombinant, human full length Casein Kinase 2α. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8807