Learn More
Eurofins Discoverx Casein Kinase 2, active, 250 µg. (14-197M)
Supplier:  Eurofins Discoverx 14197M
Encompass
Recombinant, human full length Casein Kinase 2 protein, containing an α-subunit N-terminal 6xHis tag and a β-subunit N-terminal GST tag. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8722