Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Casein Kinase 1 alpha Antibody (YA564) | Affinity purified | 34 kDa | 50 UL
SDP

Catalog No. 5000397578
Encompass_Preferred

Casein Kinase 1 alpha Antibody (YA564) is a recombinant, monoclonal IgG antibody derived from Rabbit, specifically designed for targeting Casein Kinase 1 alpha. This non-conjugated antibody is intended for research use only and is formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% Sodium azide as a preservative.

  • Rabbit-derived host
  • Recombinant, monoclonal antibody
  • Reacts with human and rat species
  • Immunogen is a synthetic peptide corresponding to human Casein Kinase 1 alpha
  • Suitable for Western blot (WB) and immunoprecipitation (IP) applications
  • Endogenous sensitivity
  • Non-conjugated
  • IgG isotype
  • Appears as a liquid
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles

Catalog No. 50-003-97578 Supplier Medchemexpress LLC Supplier No. HYP8058550UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Casein Kinase 1 alpha Antibody (YA564) is a recombinant, monoclonal IgG antibody derived from Rabbit, specifically designed for targeting Casein Kinase 1 alpha. This non-conjugated antibody is intended for research use only and is formulated in 50 mM Tris-Glycine (pH 7.4), 0.15 M NaCl, 40% Glycerol and 0.05% BSA, with 0.01% Sodium azide as a preservative.

  • Rabbit-derived host
  • Recombinant, monoclonal antibody
  • Reacts with human and rat species
  • Immunogen is a synthetic peptide corresponding to human Casein Kinase 1 alpha
  • Suitable for Western blot (WB) and immunoprecipitation (IP) applications
  • Endogenous sensitivity
  • Non-conjugated
  • IgG isotype
  • Appears as a liquid
  • Store at -20°C for 1 year
  • Avoid repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.