Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Casein kinase 1δ-IN-1 | 851871-94-8 | 99.9% | 229.26 | 1 ML
SDP

Catalog No. 5000363027
Encompass_Preferred

Casein kinase 1δ-IN-1 is an inhibitor of casein kinase 1δ (CK1δ) and can be used for the research of neurodegenerative diseases such as tauopathies. It is relevant for studies in neurological, eye or ear disease, cancer, and Alzheimer's disease.

  • Inhibits casein kinase 1δ (CK1δ)
  • Suitable for research in neurodegenerative diseases like tauopathies
  • Appears as an off-white to light brown solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 41.67 mg/mL

Catalog No. 50-003-63027 Supplier Medchemexpress LLC Supplier No. HY14811210MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Casein kinase 1δ-IN-1 is an inhibitor of casein kinase 1δ (CK1δ) and can be used for the research of neurodegenerative diseases such as tauopathies. It is relevant for studies in neurological, eye or ear disease, cancer, and Alzheimer's disease.

  • Inhibits casein kinase 1δ (CK1δ)
  • Suitable for research in neurodegenerative diseases like tauopathies
  • Appears as an off-white to light brown solid
  • Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
  • Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
  • Solubility in DMSO: 41.67 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.