Learn More
New England Biolabs, Inc. Casein Kinase II (CK2) – 50000 units
SDP
Supplier:  New England Biolabs, Inc. P6010L
Encompass
Casein Kinase II (CKII) is a constitutively active serine/threonine protein kinase composed of two 44 kDa catalytic alpha subunits and two 26 kDa regulatory beta subunits in an alpha2 beta2 configuration to form stable heterotetramers. The general recognition motif for phsophorylation by CKII is SXXE/D.
Catalog No. 50-444-2