Learn More
Enzo Life Sciences Casein kinase 2 (human), (recombinant) (10 kU)
SDP
Supplier:  Enzo Life Sciences BMLSE1240010
Encompass_Preferred
Alternative name: CK2. MW: 44 and 26 kDa subunits. Formulation: Liquid. In 20mM TRIS-HCl, pH 7.5, containing 0.35M sodium chloride, 1 mM EDTA, 2 mM DTT and 0.1% Triton X-100. Source: Produced in E. coli. Consists of 44kDa α catalytic and 26kDa ß regulatory subunits as an α2ß2 tetramer. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C. Long Term Storage: -80°C
Catalog No. NC1682766