Learn More
Medchemexpress LLC HY-111378 5mg Medchemexpress, Casein Kinase II Inhibitor IV CAS:863598-09-8 Purity:>98%
SDP
Catalog No. 501968845
Encompass_Preferred
Medchemexpress, HY-111378 5mg Casein Kinase II Inhibitor IV CAS:863598-09-8 Casein Kinase II Inhibitor IV is a small-molecule inducer of epidermal keratinocyte differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8845 Supplier Medchemexpress LLC Supplier No. HY1113785MG
Medchemexpress, HY-111378 5mg Casein Kinase II Inhibitor IV CAS:863598-09-8 Casein Kinase II Inhibitor IV is a small-molecule inducer of epidermal keratinocyte differentiation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.