Learn More
Medchemexpress LLC Casein kinase inhibitor A86 | 2079069-01-3 | 99.3% | 344.43 | C18H25FN6 | 100MG
SDP
Catalog No. 5000235155
Encompass_Preferred
Casein kinase inhibitor A86 is a potent, orally active small-molecule inhibitor of casein kinase 1α (CK1α) with reported activity against CDK7 and CDK9; it is used as a research reagent to study CK1α signaling and has shown anti-leukemic activity in preclinical studies.
  • Potent inhibitor of casein kinase 1α.
  • Also inhibits CDK7 and CDK9.
  • Shown to induce leukemia cell apoptosis in preclinical studies.
  • Available as powder and as a 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Recommended storage preserves stability for long-term use.
Catalog No. 50-002-35155 Supplier Medchemexpress LLC Supplier No. HY123955100MG
Casein kinase inhibitor A86 is a potent, orally active small-molecule inhibitor of casein kinase 1α (CK1α) with reported activity against CDK7 and CDK9; it is used as a research reagent to study CK1α signaling and has shown anti-leukemic activity in preclinical studies.
  • Potent inhibitor of casein kinase 1α.
  • Also inhibits CDK7 and CDK9.
  • Shown to induce leukemia cell apoptosis in preclinical studies.
  • Available as powder and as a 10 mM solution in DMSO.
  • High purity suitable for research applications.
  • Recommended storage preserves stability for long-term use.