Learn More
Medchemexpress LLC Casein kinase inhibitor A86 | 2079069-01-3 | 99.3% | 344.43 g/mol | C18H25FN6 | 10MM 1ML
SDP
Catalog No. 5000236862
Encompass_Preferred
Casein kinase inhibitor A86 is a potent small-molecule inhibitor of casein kinase 1α (CK1α) with reported activity against CDK7 and CDK9. Supplied as a 10 mM solution in DMSO, it is intended for biochemical and cell-based research applications, including studies of apoptosis and leukemia-related signaling pathways.
  • Potent CK1α inhibitor with activity against CDK7 and CDK9.
  • Supplied as a 10 mM solution in DMSO for ready use in assays.
  • High purity suitable for research: 99.3%.
  • Molecular formula C18H25FN6; molecular weight 344.43 g/mol.
  • Volume 1 mL, convenient for small-scale experiments.
  • For research use only; not for human therapeutic applications.
Catalog No. 50-002-36862 Supplier Medchemexpress LLC Supplier No. HY12395510MM 1ML
Casein kinase inhibitor A86 is a potent small-molecule inhibitor of casein kinase 1α (CK1α) with reported activity against CDK7 and CDK9. Supplied as a 10 mM solution in DMSO, it is intended for biochemical and cell-based research applications, including studies of apoptosis and leukemia-related signaling pathways.
  • Potent CK1α inhibitor with activity against CDK7 and CDK9.
  • Supplied as a 10 mM solution in DMSO for ready use in assays.
  • High purity suitable for research: 99.3%.
  • Molecular formula C18H25FN6; molecular weight 344.43 g/mol.
  • Volume 1 mL, convenient for small-scale experiments.
  • For research use only; not for human therapeutic applications.