Learn More
Echelon Biosciences Research Labs Casein Kinase Substrate - 1 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 871561MG
Encompass_Preferred
Casein Kinase Substrate - 1mg , Arg-Arg-Arg-Glu-Glu-Glu-Thr-Glu-Glu-Glu-OH Category/Application: Enzyme Substrates This item is non returnable
Catalog No. 50-190-9012