Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Casein Peptone

Catalog No. aah2655736
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2655736 500 g
AAH2655722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2655736 Supplier Thermo Scientific Chemicals Supplier No. H2655736
Only null left
Add to Cart

Casein peptone is an enzymatic derivative of casein, CAS # 9000-71-9, the protein found in milk and animal tissues.

Casein peptone is used in nutrient media for growing bacteria and fungi. It is employed in antibiotics and enzyme productions. It is also essential for analytical microbiology and industrial fermentation process. It is also used in bacterial diagnostics and in agro-food, cosmetic and pharmaceutical fermentation for production of biomass.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Casein peptone is commonly used in the preparation of culture media in microbiology

• Casein peptone is an enzymatically prepared hydrolysate of casein

• Casein peptone is an opioid peptide and a rich source of amino acids

Application

• Supports the growth of a plethora of organisms in culture, particularly in serum-free media

• Enzymatic digest of casein peptone can provide active peptides for research on opioid signaling

TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Packaging Type Glass Bottle
Quantity 500 g
Sensitivity Hygroscopic

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.