Learn More
NEOGEN Casein Peptone Type I, 5 kg
SDP
Supplier:  NEOGEN 700003313
Encompass_Preferred
Casein Peptone Type I is an enzymatic hydrolysate of casein for use in preparing microbiological culture media in a laboratory setting and is not intended for use in the diagnosis of disease or other conditions in humans.Casein Peptone Type I is an enzymatic hydrolysate of casein, rich in peptones and amino acids (including tryptophane). This peptone can be utilized by most bacteria as a growth substrate.For more information visit https://www.neogen.com/solutions/Microbiology/casein-peptone-type-1
Catalog No. 50-201-5499