Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh
Used as cheese, food additive and binder for safety matches.
Supplier: MP Biomedicals™ 0290452025
Description
- Phosphoprotein
- Extracted from bovine milk
- Used as food source
- CAS: 9000-71-9

Specifications
Casein | |
Powder | |
25 lb. |
Yellow | |
Used as cheese, food additive and binder for safety matches | |
Store at room temperature (15°C to 30°C) |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction