Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein, Vitamin Tested, 30 Mesh

Catalog No. MP290452080 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 lb.
5 lb.
25 lb.
100 lb.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP290452080 100 lb.
ICN90452001 1 lb.
ICN90452005 5 lb.
MP290452025 25 lb.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP290452080 Supplier MP Biomedicals™ Supplier No. 0290452080
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Used as cheese, food additive and binder for safety matches.

  • Phosphoprotein
  • Extracted from bovine milk
  • Used as food source
  • CAS: 9000-71-9
TRUSTED_SUSTAINABILITY

Specifications

Product Type Casein
Color Yellow
Format Powder
For Use With (Application) Used as cheese, food additive and binder for safety matches
Quantity 100 lb.
Storage Requirements Store at room temperature (15°C to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.