Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Casein Sodium Salt - New Zealand Origin

Catalog No. MP029028975 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP029028975 500 g
MP290289701 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP029028975 Supplier MP Biomedicals™ Supplier No. 02902897.5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS Number: 9005-46-3

Casein protein has numerous experimental applications including use as a blocking agent in immunochemistry, recovery of enzyme activity from SDS extracted samples, and as a substrate for protease and kinase assays.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Not Autoclavable
Product Type Animal Diet
Disposable Single-use
Quantity 500 g
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 9 in.
Length (Metric) 0.2286 m
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.