Learn More
Vector Laboratories Casein Solution 10X 250 ml
SDP
Catalog No. NC9965082
Encompass
Casein Solution is a general blocking agent for immunohistochemistry assays, nucleic acid and protein blotting, and other applications. In this product, it is supplied as a 10x concentrate.
Catalog No. NC9965082 Supplier Vector Laboratories Supplier No. SP5020250
Casein Solution is a general blocking agent for immunohistochemistry assays, nucleic acid and protein blotting, and other applications. In this product, it is supplied as a 10x concentrate.